Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ MyLab™ Custom Kit - Forensics Blood Typing
SDPSCI_ED

Catalog No. S06464 Shop All EDVOTEK Products
Change view
Click to view available options
MyLab™ Custom Kit - Forensics Blood Typing
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S06464 MyLab™ Custom Kit - Forensics Blood Typing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06464 Supplier EDVOTEK™ Supplier No. 1191

Are you planning a virtual curriculum or looking for experiments ideal for distance learning? With the MyLab™ Custom Kits, you can easily plan your curriculum. Each kit comes with instructions and materials for 2-3 runs of each experiment.

  • Learn about basic forensic detection techniques
  • Swab simulated crime scene samples to detect blood left at the scene
  • Analyze the crime scene samples to determine the suspect's blood type
  • Understand presumptive versus confirmatory testing

Specifications

Description Forensics Blood Typing
Grade 9 to 15
Age 14 to 20
For Use With (Application) At Home Education
Includes Graduated Transfer Pipettes, Microcentrifuge Tubes, Simulated Blood Samples, Cotton Swabs, Hydrogen Peroxide Solution, Phenolphthalein Stock Solution, Evidence Bag, Red Dye Concentrate, Microtiter Strip, Anti-A and Anti-B Serum
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.