Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ MyLab™ Custom Kit - How Do We Keep Our Food Safe?
SDPSCI_ED

Catalog No. S06472 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
9 to 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S06472 9 to 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06472 Supplier EDVOTEK™ Supplier No. 1213

missing translation for 'validMainframeMsg'

Are you planning a virtual curriculum or looking for experiments ideal for distance learning? With the MyLab™ Custom Kits, you can easily plan your curriculum. Each kit comes with instructions and materials for 2-3 runs of each experiment.

Bacteria grows and multiplies rapidly unless kept in check by good sanitation, food safety, and personal hygiene. Many procedures are routinely used in food safety that include cooking, chilling, cleaning, and avoiding cross-contamination. In this experiment, milk will be used as a growth medium for bacteria. A blue dye will be added that turns colorless when bacteria grow in it. The impact of cooking, chilling, and cross-contamination will be tested to determine bacterial growth.

  • Learn how biotechnologies and the 4Cs protect food from pathogenic bacteria
  • Explore how temperature affects bacteria growth
  • Open ended experiment lets students design and test additional variables
  • Includes indicator dye, pH paper, acetic acid, sodium bicarbonate, tubes, pipettes

Specifications

Grade 9 to 15
Age 14 to 20
For Use With (Application) At Home Education
Includes Indicator Dye, pH Paper, Acetic Acid, Sodium Bicarbonate, Tubes, Pipettes
Quantity 1 Kit
Description How Do We Keep Our Food Safe?
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.