Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ MyLab™ Custom Kit - Simulation of COVID-19 Antibody Test
SDPSCI_ED

Catalog No. S06462 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
2 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S06462 2 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06462 Supplier EDVOTEK™ Supplier No. 1219
Add to Cart
Add to Cart

Are you planning a virtual curriculum or looking for experiments ideal for distance learning? With the MyLab™ Custom Kits, you can easily plan your curriculum. Each kit comes with instructions and materials for 2-3 runs of each experiment.

  • Learn about antibody-based detection techniques
  • Perform an ELISA at home
  • Understand current testing techniques for COVID-19 through a simulated experiment
  • Includes controls, antibodies, test strips, detection substrate, transfer pipettes

Specifications

Grade 9 to 15
Age 14 to 20
For Use With (Application) At Home Education
Includes Controls, Antibodies, Test Strips, Detection Substrate, Transfer Pipettes
Quantity 1 Kit
Class Size 2 Students
Product Type ELISA Kit
Description Simulation of COVID-19 Antibody Test
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.