Learn More
Premier Lab Supply Mylar 6.0μm (0.24mil) Sheets – PN# TF-160-345
SDP
Catalog No. NC9732680
Encompass_Preferred
Mylar film 6.0μm (0.24mil) Sheets with Dimensions: 3” x 3” (76 x 76mm) (PN# TF-160-345) – Mylar has excellent tinsel strength and chemical resistance, and is well-suited for light elements in thinner gauges. 500 per box.
Catalog No. NC9732680 Supplier Premier Lab Supply Supplier No. TF160345
Mylar film 6.0μm (0.24mil) Sheets with Dimensions: 3” x 3” (76 x 76mm) (PN# TF-160-345) – Mylar has excellent tinsel strength and chemical resistance, and is well-suited for light elements in thinner gauges. 500 per box.