Learn More
Premier Lab Supply Mylar 3.5μm (0.14mil) Continuous Roll – PN# TF-135
SDP
Catalog No. NC1921860
Encompass_Preferred
Mylar film 3.5μm (0.14mil) continuous roll with Dimensions: 3” x 300’ (76mm x 91.4m) (PN# TF-135) – Mylar has excellent tinsel strength and chemical resistance, and is well-suited for light elements in thinner gauges.
Catalog No. NC1921860 Supplier Premier Lab Supply Supplier No. TF135
Mylar film 3.5μm (0.14mil) continuous roll with Dimensions: 3” x 300’ (76mm x 91.4m) (PN# TF-135) – Mylar has excellent tinsel strength and chemical resistance, and is well-suited for light elements in thinner gauges.