Learn More
Premier Lab Supply Mylar 3.5μm (0.14mil) Pre-Cut Circles, 2.5” Diameter – PN# TF-135-255
SDP
Catalog No. NC1680871
Encompass_Preferred
Mylar film 3.5μm (0.14mil) cut into easy-to-use circles with 2.5” Diameter (PN# TF-135-255) – Mylar has excellent tinsel strength and chemical resistance, and is well-suited for light elements in thinner gauges. 500 per box
Catalog No. NC1680871 Supplier Premier Lab Supply Supplier No. TF135255
Mylar film 3.5μm (0.14mil) cut into easy-to-use circles with 2.5” Diameter (PN# TF-135-255) – Mylar has excellent tinsel strength and chemical resistance, and is well-suited for light elements in thinner gauges. 500 per box