Learn More
Premier Lab Supply Mylar 6.0μm (0.24mil) Continuous Roll – PN# TF-160
SDP
Catalog No. NC0149135
Encompass_Preferred
Mylar film 6.0μm (0.24mil) continuous roll with Dimensions: 3” x 300’ (76mm x 91.4m) (PN# TF-160) – Mylar has excellent tinsel strength and chemical resistance, and is well-suited for light elements in thinner gauges.
Catalog No. NC0149135 Supplier Premier Lab Supply Supplier No. TF160
Mylar film 6.0μm (0.24mil) continuous roll with Dimensions: 3” x 300’ (76mm x 91.4m) (PN# TF-160) – Mylar has excellent tinsel strength and chemical resistance, and is well-suited for light elements in thinner gauges.