Learn More
Medchemexpress LLC HY-136446 50mg , MYLS22 CAS:306959-01-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13644650MG
Encompass_Preferred
HY-136446 50mg MYLS22 CAS: 306959-01-3 MYLS22 is a first-in-class and selective optic atrophy 1 (OPA1) inhibitor. MYLS22 can target endothelial OPA1 to curtail tumor growth and inhibits angiogenesis by impinging on NFkB activity and on angiogenic gene expression. MYLS22 can inhibit mitochondrial fusion and exacerbate the mitochondrial damage. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9711