Learn More
Medchemexpress LLC HY-136446 5mg Medchemexpress, MYLS22 CAS:306959-01-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1364465MG
Encompass_Preferred
Medchemexpress, HY-136446 5mg MYLS22 CAS:306959-01-3 MYLS22 is a first-in-class and selective optic atrophy 1 (OPA1) inhibitor. MYLS22 can target endothelial OPA1 to curtail tumor growth and inhibits angiogenesis by impinging on NFkB activity and on angiogenic gene expression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0440
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.