Learn More
Apexbio Technology LLC myo-Inositol 87-89-8 5g
SDP
Catalog No. 502240395
Encompass_Preferred
myo-Inositol (CAS 87-89-8) is a six-carbon cyclic polyol that serves as a metabolic precursor and structural component in eukaryotic cells It is incorporated into membrane phospholipids and forms the basis for glycosylphosphatidylinositol (GPI) anchors which tether glycoproteins to the cell surface Additionally myo-Inositol is involved in the generation of inositol phosphate derivatives that function as intracellular second messengers mediating signal transduction pathways It is widely utilized in biomedical research to study membrane dynamics cell signaling and glycosylation processes
This item is not returnable. View return policy
Catalog No. 50-224-0395 Supplier Apexbio Technology LLC Supplier No. B79035000
myo-Inositol (CAS 87-89-8) is a six-carbon cyclic polyol that serves as a metabolic precursor and structural component in eukaryotic cells It is incorporated into membrane phospholipids and forms the basis for glycosylphosphatidylinositol (GPI) anchors which tether glycoproteins to the cell surface Additionally myo-Inositol is involved in the generation of inositol phosphate derivatives that function as intracellular second messengers mediating signal transduction pathways It is widely utilized in biomedical research to study membrane dynamics cell signaling and glycosylation processes