Learn More
Medchemexpress LLC Myo-inositol-d6 | 68922-44-1 | MFCD01074110 | 99.9% | 186.19 | C6H6D6O6 | 10 MG
SDP
Catalog No. 5000298924
Encompass_Preferred
Deuterium-labeled myo-inositol (i-Inositol-d6) is a fully deuterated isotopologue of myo-inositol provided in small, pre-weighed quantities for analytical and biochemical use. It is intended for applications that require a stable-isotope standard or tracer for mass spectrometry and metabolic studies.
  • Stable isotope-labeled (D6) for tracer and quantitation studies.
  • High purity (99.89%) suitable for LC-MS and GC-MS reference standards.
  • Convenient small, pre-weighed packages for analytical workflows.
  • Molecular formula C6H6D6O6 and molecular weight 186.19 for precise calculation and reporting.
  • Common applications include internal standard use, metabolic tracing, and analytical reference material.
Catalog No. 50-002-98924 Supplier Medchemexpress LLC Supplier No. HYB1411S10MG
Deuterium-labeled myo-inositol (i-Inositol-d6) is a fully deuterated isotopologue of myo-inositol provided in small, pre-weighed quantities for analytical and biochemical use. It is intended for applications that require a stable-isotope standard or tracer for mass spectrometry and metabolic studies.
  • Stable isotope-labeled (D6) for tracer and quantitation studies.
  • High purity (99.89%) suitable for LC-MS and GC-MS reference standards.
  • Convenient small, pre-weighed packages for analytical workflows.
  • Molecular formula C6H6D6O6 and molecular weight 186.19 for precise calculation and reporting.
  • Common applications include internal standard use, metabolic tracing, and analytical reference material.