Learn More
Alkali Scientific Myo Inositol [I Inositol], 500 G
SDP
Supplier:  Alkali Scientific MYO004CP
Encompass_Preferred
Myo inositol is a naturally occurring compound used in laboratory applications as a vitamin-like growth factor for cells. This 500g reagent is essential for supporting the growth and maintenance of various mammalian cell lines, particularly those used in metabolic and genetic research. Myo inositol is a key player in regulating cell signaling and membrane formation, making it vital for studies on cellular development and health. It is commonly included in cell culture media for its role in enhancing cell survival, proliferation, and differentiation. This compound is also used in research on metabolic disorders, such as diabetes, and in studies related to brain and nerve function.
Catalog No. 50-006-61455