Learn More
ABclonal Technology MYO19 Rabbit mAb
SDP
Catalog No. 5000007957
Encompass_Preferred
Enables actin binding activity Involved in regulation of cytokinesis and regulation of mitochondrial fission Acts upstream of or within mitochondrion migration along actin filament Located in cytosol and mitochondrial outer membrane
Catalog No. 50-000-07957 Supplier ABclonal Technology Supplier No. A2093720UL
Enables actin binding activity Involved in regulation of cytokinesis and regulation of mitochondrial fission Acts upstream of or within mitochondrion migration along actin filament Located in cytosol and mitochondrial outer membrane