Learn More
Bioworld MYOBACTERIUM TUBERCULOSIS D
Supplier:  Bioworld 220607371
Encompass_Preferred
MYOBACTERIUM TUBERCULOSIS DNAK (HSP70), (RECOMBINANT): Recombinant Mycobacterium Tuberculosis Heat Shock Protein 70kD Source: E.Coli Description : Recombinant Mycobacterium Tuberculosis Heat Shock Protein is produced in E.Coli . Myco...
Catalog No. 50-253-835