Learn More
Aviva Systems Biology Corp Myocd Antibody - N-terminal region (ARP50817_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50817_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Myocd. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: BSAC2A, Srfcp, MyocdApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-543