Learn More
STEMCELL Technologies MyoCult™ Differentiation Kit (Human), Size: 200 mL
SDP
Catalog No. 501974368
Encompass
MyoCult™ Differentiation Kit (Human) includes MyoCult™ Differentiation Medium and Animal Component-Free (ACF) Cell Attachment Substrate. MyoCult™ Differentiation Medium is a serum-based medium specifically formulated for the in vitro differentiation of human skeletal muscle progenitor cells (myoblasts) into myotubes. MyoCult™ Differentiation Medium is designed for use with the ACF Cell Attachment Substrate, which has been specifically tested for its ability to support optimal adherence and myotube morphology. This medium is suitable for the differentiation of myogenic progenitor cells previously expanded in serum-containing or serum-free media (e.g. using the MyoCult™-SF Expansion Kit #05980).CollPlant (https://collplant.com/) is the manufacturer of the rhCollagen component of the cell attachment substrate.
Catalog No. 50-197-4368 Supplier STEMCELL Technologies Supplier No. 05965
MyoCult™ Differentiation Kit (Human) includes MyoCult™ Differentiation Medium and Animal Component-Free (ACF) Cell Attachment Substrate. MyoCult™ Differentiation Medium is a serum-based medium specifically formulated for the in vitro differentiation of human skeletal muscle progenitor cells (myoblasts) into myotubes. MyoCult™ Differentiation Medium is designed for use with the ACF Cell Attachment Substrate, which has been specifically tested for its ability to support optimal adherence and myotube morphology. This medium is suitable for the differentiation of myogenic progenitor cells previously expanded in serum-containing or serum-free media (e.g. using the MyoCult™-SF Expansion Kit #05980).CollPlant (https://collplant.com/) is the manufacturer of the rhCollagen component of the cell attachment substrate.