Learn More
STEMCELL Technologies MyoCult Expansion 10X Supplement (Mouse), Size: 10 mL
SDP
Catalog No. 502390314
Encompass
MyoCult Expansion 10X Supplement (Mouse) when combined with DMEM/F-12 to prepare MyoCult Expansion Medium is for the culture of mouse skeletal muscle progenitor cells (satellite cells). MyoCult Expansion Medium has been optimized for the expansion of mouse skeletal muscle satellite cells isolated by fluorescence-activated cell sorting (FACS) as well as for culturing single isolated myofibers grown in suspension. Satellite cells cultured using MyoCult Expansion Medium can be differentiated into multinucleated myotubes
Catalog No. 50-239-0314 Supplier STEMCELL Technologies Supplier No. 05985
MyoCult Expansion 10X Supplement (Mouse) when combined with DMEM/F-12 to prepare MyoCult Expansion Medium is for the culture of mouse skeletal muscle progenitor cells (satellite cells). MyoCult Expansion Medium has been optimized for the expansion of mouse skeletal muscle satellite cells isolated by fluorescence-activated cell sorting (FACS) as well as for culturing single isolated myofibers grown in suspension. Satellite cells cultured using MyoCult Expansion Medium can be differentiated into multinucleated myotubes