Learn More
STEMCELL Technologies MyoCult-SF Attachment Substrate, Size: 100 ug
SDP
Catalog No. 502390428
Encompass
Serum-free attachment substrate for the derivation and expansion of human skeletal muscle progenitor cells (myoblasts)
Catalog No. 50-239-0428 Supplier STEMCELL Technologies Supplier No. 05983
Serum-free attachment substrate for the derivation and expansion of human skeletal muscle progenitor cells (myoblasts)