Learn More
STEMCELL Technologies MyoCult-SF Expansion Supplement Kit (Human), Size: 1 Kit
SDP
Catalog No. 502390209
Encompass
Serum-free supplement and attachment substrate for the derivation and expansion of human skeletal muscle progenitor cells (myoblasts)
Catalog No. 50-239-0209 Supplier STEMCELL Technologies Supplier No. 05980
Serum-free supplement and attachment substrate for the derivation and expansion of human skeletal muscle progenitor cells (myoblasts)