Learn More
Promocell GmbH PromoCell Myocyte Basal Medium 500 ml
Catalog No. 50306210
Encompass
The PromoCell Myocyte Growth Medium is a low serum (5% V/V) medium supplemented with well chosen hormones and growth factors. It was developed for the optimal expansion and maintenance of primary cardiac myocytes in culture.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for murine and primary rat cardiac myocytes.| Media supplement | Final supplement concentration|| Fetal Calf Serum | 0.05 ml / ml || Epidermal Growth Factor (recombinant human) | 0.5 ng / ml || Basic Fibroblast Growth Factor (recombinant human) | 2 ng / ml || Insulin (recombinant human) | 5 µg / ml |
Catalog No. 50-306-210 Supplier Promocell GmbH Supplier No. C22270
The PromoCell Myocyte Growth Medium is a low serum (5% V/V) medium supplemented with well chosen hormones and growth factors. It was developed for the optimal expansion and maintenance of primary cardiac myocytes in culture.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for murine and primary rat cardiac myocytes.| Media supplement | Final supplement concentration|| Fetal Calf Serum | 0.05 ml / ml || Epidermal Growth Factor (recombinant human) | 0.5 ng / ml || Basic Fibroblast Growth Factor (recombinant human) | 2 ng / ml || Insulin (recombinant human) | 5 µg / ml |