Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4654MSM3P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: 5.8A + MYD712. Clonality: Monoclonal. Host: Mouse. Isotype: IgG. Positive Control: Rhabdomyosarcoma. Cellular Staining: Nuclear. Only nuclear staining should be considered as evidence of skeletal muscle differentiation. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant mouse MyoD1 protein (5.8A); Recombinant human MyoD1 protein (MYD712). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 45kDa.Chromosome: 11p15.1. Gene Symbol: MYOD1. Gene ID: 4654. Ensemble: ENSG00000129152. Uniprot: NP_002469.
Catalog No. 50-188-4991