Learn More
Aviva Systems Biology Corp MYOD1 Antibody - middle region (ARP38158_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38158_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MYOD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MYOD1. Alternate names: MYF3, MYOD, PUM, bHLHc1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56545