Learn More
Biocare Medical, LLC Myogenin Antibody - Concentrate, 0.1 ml
SDP
Supplier:  Biocare Medical, LLC CM115A
Encompass_Preferred
Myogenin is a member of a family of myogenic genes that also includes MyoD. These genes encode a set of transcription factors that are essential for muscle development. Expression of myogenin is restricted to cells of skeletal muscle origin. Myogenin antibody has been shown to label human myogenin and label neonatal mouse, rat and cat tissues. Staining has also been found in myoblasts from human fetal limbs. No reactivity was found in adult skeletal muscle. Myogenin has been observed to stain the vast majority of rhabdomyosarcomas and Wilm tumors. No activity was observed in Ewing sarcoma/peripheral primitive neuroectodermal tumor, or in neuroblastomas.
Catalog No. 50-825-23