Learn More
Diagnostic Biosystems MYOGENIN Myogenin
Catalog No. 502407608
Encompass_Preferred
This antibody is specific to 34 kDa protein known as myogenin. Myogenin is a member of family of myogenic regulatory genes which includes MyoD, myf5 and MRF4. These genes encode a set of transcription factors which are essential for muscle development. Expression of myogenin is restricted to cells of skeletal muscle origin.
Catalog No. 50-240-7608 Supplier Diagnostic Biosystems Supplier No. PDM158
This antibody is specific to 34 kDa protein known as myogenin. Myogenin is a member of family of myogenic regulatory genes which includes MyoD, myf5 and MRF4. These genes encode a set of transcription factors which are essential for muscle development. Expression of myogenin is restricted to cells of skeletal muscle origin.