Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4656MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: MGN185. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Rh-30 cells. Skeletal muscle or rhabdomyosarcoma. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Human myogenin recombinant protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 34kDa.Chromosome: 1q32.1. Gene Symbol: MYOG. Gene ID: 4656. Ensemble: ENSG00000122180. Uniprot: NP_002470.
Catalog No. 50-188-4992