Learn More
Genscript Corporation Antibody Protein Proteins Proteins 10 UG 1/EA
Supplier:  Genscript Corporation Z0331910
Encompass
Myoglobin a member of the globin family of proteins is a cytosolic oxygen-binding protein that regulates the storage and diffusion of oxygen within myocytes. The largest expression of myoglobin is in skeletal and cardiac muscle. Myoglobin exhibits various functions in relation to the muscular oxygen supply such as oxygen storage facilitated diffusion and myoglobin-mediated oxidative phosphorylation. Myoglobin is the primary oxygen-carrying pigment of muscle tissues. High concentrations of myoglobin in muscle cells allow organisms to hold their breath for a longer period of time. Diving mammals such as whales and seals have muscles with a particularly high abundance of myoglobin. Myoglobin is found in Type I Type II A and Type II B muscle; however several studies indicate myoglobinis not found in smooth muscle.
Catalog No. 50-252-9662