Learn More
Medchemexpress LLC Myoglobin Mouse P. 10ug
SDP
Catalog No. 5000195941
Encompass_Preferred
Recombinant mouse myoglobin expressed in Pichia pastoris is supplied as a lyophilized, His-tagged full-length protein intended for research use in biochemical, structural, and antibody validation applications. The product is formulated for stability and storage and is suitable for low-volume assays and detection workflows.
  • Full-length mature myoglobin suitable for functional and structural studies.
  • Expressed in Pichia pastoris to support eukaryotic processing.
  • N-terminal 6xHis tag for straightforward purification and detection.
  • Lyophilized formulation for extended shelf life and convenient transport.
  • Formulated with trehalose to improve stability upon reconstitution.
  • High purity (>90%) as assessed by reducing SDS-PAGE.
  • Available in small vials for low-volume experiments and assays.
Catalog No. 50-001-95941 Supplier Medchemexpress LLC Supplier No. HYP7175410UG
Recombinant mouse myoglobin expressed in Pichia pastoris is supplied as a lyophilized, His-tagged full-length protein intended for research use in biochemical, structural, and antibody validation applications. The product is formulated for stability and storage and is suitable for low-volume assays and detection workflows.
  • Full-length mature myoglobin suitable for functional and structural studies.
  • Expressed in Pichia pastoris to support eukaryotic processing.
  • N-terminal 6xHis tag for straightforward purification and detection.
  • Lyophilized formulation for extended shelf life and convenient transport.
  • Formulated with trehalose to improve stability upon reconstitution.
  • High purity (>90%) as assessed by reducing SDS-PAGE.
  • Available in small vials for low-volume experiments and assays.