Learn More
Medchemexpress LLC Myoglobin Mouse P. 50ug
SDP
Catalog No. 5000196090
Encompass_Preferred
Recombinant mouse myoglobin produced in Pichia pastoris and supplied lyophilized with N-terminal 6xHis and C-terminal Myc tags. The preparation is >90% pure by reducing SDS-PAGE, formulated with Tris-HCl, NaCl, and trehalose for stability, and intended for laboratory research applications.
  • High purity suitable for biochemical assays.
  • Expressed in Pichia pastoris for eukaryotic folding.
  • His and Myc tags facilitate purification and detection.
  • Lyophilized formulation enhances shelf life and transport.
  • Formulated with trehalose to improve stability on storage.
  • Suitable for binding studies and analytical standards.
Catalog No. 50-001-96090 Supplier Medchemexpress LLC Supplier No. HYP7175450UG
Recombinant mouse myoglobin produced in Pichia pastoris and supplied lyophilized with N-terminal 6xHis and C-terminal Myc tags. The preparation is >90% pure by reducing SDS-PAGE, formulated with Tris-HCl, NaCl, and trehalose for stability, and intended for laboratory research applications.
  • High purity suitable for biochemical assays.
  • Expressed in Pichia pastoris for eukaryotic folding.
  • His and Myc tags facilitate purification and detection.
  • Lyophilized formulation enhances shelf life and transport.
  • Formulated with trehalose to improve stability on storage.
  • Suitable for binding studies and analytical standards.