Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Myoglobin Mouse P. 5ug
SDP

Catalog No. 5000196260
Encompass_Preferred

Recombinant mouse myoglobin expressed in Pichia pastoris and supplied as a purified, tagged protein for use in biochemical assays, structural studies, and antibody validation. The product includes C-Myc and N-terminal 6xHis tags to facilitate purification and detection, and is provided with documentation including a datasheet, certificate of analysis, and SDS for quality assurance.

  • Recombinant myoglobin produced in Pichia pastoris.
  • C-Myc and N-terminal 6xHis tags facilitate purification and detection.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Approximate molecular weight of 21 kDa.
  • Supplied in small quantities suitable as assay controls or positive controls.
  • Store at -20°C for long-term storage; stable at 4°C for short term after reconstitution.
  • Datasheet, COA, and SDS available for quality and handling information.

Catalog No. 50-001-96260 Supplier Medchemexpress LLC Supplier No. HYP717545UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse myoglobin expressed in Pichia pastoris and supplied as a purified, tagged protein for use in biochemical assays, structural studies, and antibody validation. The product includes C-Myc and N-terminal 6xHis tags to facilitate purification and detection, and is provided with documentation including a datasheet, certificate of analysis, and SDS for quality assurance.

  • Recombinant myoglobin produced in Pichia pastoris.
  • C-Myc and N-terminal 6xHis tags facilitate purification and detection.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Approximate molecular weight of 21 kDa.
  • Supplied in small quantities suitable as assay controls or positive controls.
  • Store at -20°C for long-term storage; stable at 4°C for short term after reconstitution.
  • Datasheet, COA, and SDS available for quality and handling information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.