Learn More
MyBioSource Myoglobin Protien 1mg
SDP
Supplier:  MyBioSource MBS1731651 MG
Encompass_Preferred
Myoglobin from MyBioSource is a 17.8 kDa protein purified from equine skeletal muscle. It is supplied as a lyophilized powder and is >95% pure as determined by SDS-PAGE. Myoglobin is an oxygen-binding protein found in muscle tissue that helps store oxygen in muscle cells. This product can be used in activity assays and immunological detection. Myoglobin from MyBioSource is an important research reagent for studying muscle physiology and oxygen transport.
Catalog No. NC2751008