Learn More
Aviva Systems Biology Corp MYOM1 Antibody - C-terminal region (ARP65177_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65177_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MYOM1. It was validated on Western Blot. . Alternate names: MGC134946, MGC134947, SKELEMINApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-288-451