Learn More
Medchemexpress LLC MyoMed 205 | 2614161-13-4 | 98.0% | 491.56 g/mol | C26H25N3O5S | 10 MG
SDP
Catalog No. 5000288996
Encompass_Preferred
MyoMed 205 is a small-molecule inhibitor of MuRF1 supplied as a white to off-white solid for research use. It reduces ubiquitination and proteasomal degradation of muscle proteins and has been used in preclinical studies to prevent early diaphragmatic systolic dysfunction and muscle atrophy. Recommended handling includes storage of the powder at low temperature and preparation of solutions in DMSO with warming as needed.
  • Small-molecule MuRF1 inhibitor for research use.
  • White to off-white solid with high reported purity.
  • Soluble in DMSO at up to 25 mg/mL with sonication and warming.
  • Formulatable for in vivo dosing in 10% DMSO/90% corn oil at ≥ 2.5 mg/mL.
  • Store powder at -20°C for long-term stability.
Catalog No. 50-002-88996 Supplier Medchemexpress LLC Supplier No. HY16402710MG
MyoMed 205 is a small-molecule inhibitor of MuRF1 supplied as a white to off-white solid for research use. It reduces ubiquitination and proteasomal degradation of muscle proteins and has been used in preclinical studies to prevent early diaphragmatic systolic dysfunction and muscle atrophy. Recommended handling includes storage of the powder at low temperature and preparation of solutions in DMSO with warming as needed.
  • Small-molecule MuRF1 inhibitor for research use.
  • White to off-white solid with high reported purity.
  • Soluble in DMSO at up to 25 mg/mL with sonication and warming.
  • Formulatable for in vivo dosing in 10% DMSO/90% corn oil at ≥ 2.5 mg/mL.
  • Store powder at -20°C for long-term stability.