Learn More
Cayman Chemical MyomodulIn AAplysIaLymnae 10mg
SDP
Catalog No. 502945694
Encompass_Preferred
A gastropod neuropeptide; potentiates motor neuron-elicited contractions of the A. californica buccal mass ARC; increases the relaxation rate of electrically evoked muscle contractions in L. stagnalis penis retractor muscle
Catalog No. 50-294-5694 Supplier Cayman Chemical Supplier No. 4015110 MG
A gastropod neuropeptide; potentiates motor neuron-elicited contractions of the A. californica buccal mass ARC; increases the relaxation rate of electrically evoked muscle contractions in L. stagnalis penis retractor muscle