Learn More
Medchemexpress LLC HY-W008956 5mg Medchemexpress, Myoseverin CAS:267402-71-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW0089565MG
Encompass_Preferred
Medchemexpress, HY-W008956 5mg Myoseverin CAS:267402-71-1 Myoseverina, a microtubule-binding molecule, induces the reversible fission of multinucleated myotubes into mononucleated fragments. Myoseverina affects the expression of a variety of growth factor, immunomodulatory, extracellular matrix-remodeling, and stress response genes, consistent with the activation of pathways involved in wound healing and tissue regeneration. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3264