Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyvinyl Alcohol, Hydrolyzed, Reagent, 99-100%, Spectrum™ Chemical
SDP

Catalog No. 18605401 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-605-401 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-605-401 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PO141250GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PO141, 9002-89-5, (C2H4O)n

Spectrum™ Chemical Polyvinyl Alcohol, 99-100 Percent, Hydrolyzed, Reagent, also known as PVA or PVAI, is a synthetic polymer used in textiles, papermaking, and several types of coatings. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 9002-89-5
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 44.05
MDL Number MFCD00081922
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
IUPAC Name Ethenol
SMILES OC(-*)C-*

Specifications

CAS Max % 1
Quantity 250 g
Packaging Poly Bottle
Viscosity 24-32 cps
Percent Purity 99 to 100%
Grade Reagent
Chemical Name or Material Polyvinyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.