Learn More
Biogenex Laboratories Myosin H. Chain : SMMS.1 : 6mL
SDP
Supplier:  Biogenex Laboratories AM3315M
Encompass_Preferred
SMMS-1 is approximately 204 kD and is considered to be the marker forsmooth muscle cell phenotypes. It has been designed for specificlocalization of both vascular and visceral smooth muscle. Monoclonalantibody to smooth muscle myosin heavy chains in combination withmonoclonal antibodies to calponin and heavy caldesmon may be used tostudy the differences between benign, in-situ lesions and invasivecarcinomas.
Catalog No. 50-197-8838