Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Myosin Heavy Chain Antibody (YA5348) | 220 kDa | 50 UL
SDP

Catalog No. 5000446228
Encompass_Preferred

Myosin Heavy Chain Antibody (YA5348) is a mouse-derived, non-conjugated monoclonal antibody targeting Myosin Heavy Chain. Myosin heavy chain is a non-regulatory myosin light chain essential for the proper formation and/or maintenance of myofibers, thereby ensuring appropriate muscle function.

  • Derived from mouse
  • Non-conjugated antibody
  • Reactive with human, mouse, rat, fruit fly, and nematode samples
  • Supplied in PBS, pH 7.4, with 0.5% BSA, 0.02% sodium azide, and 50% glycerol
  • Purified by affinity chromatography
  • Store at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Appears as a liquid
  • Suitable for ICC/IF applications at a 1:100 dilution
  • Ships with blue ice

Catalog No. 50-004-46228 Supplier Medchemexpress LLC Supplier No. HYP8565650UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Myosin Heavy Chain Antibody (YA5348) is a mouse-derived, non-conjugated monoclonal antibody targeting Myosin Heavy Chain. Myosin heavy chain is a non-regulatory myosin light chain essential for the proper formation and/or maintenance of myofibers, thereby ensuring appropriate muscle function.

  • Derived from mouse
  • Non-conjugated antibody
  • Reactive with human, mouse, rat, fruit fly, and nematode samples
  • Supplied in PBS, pH 7.4, with 0.5% BSA, 0.02% sodium azide, and 50% glycerol
  • Purified by affinity chromatography
  • Store at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Appears as a liquid
  • Suitable for ICC/IF applications at a 1:100 dilution
  • Ships with blue ice

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.