Learn More
Enzo Life Sciences Myosin (human) monoclonal antibody (F36.5B9) (1ml)
SDP
Supplier:  Enzo Life Sciences ALXBC1040SL001
Encompass_Preferred
Clone: F36.5B9. Isotype: IgG2a. Immunogen: Human ventricular myosin. Species reactivity: Human. Specificity: Recognizes the S1/S2 junction of human ventricular myosin heavy chain β. Applications: IHC (FS), WB. Formulation: Liquid. In sterile filtered supernatant with 0.09% sodium azide.
Catalog No. 50-201-0166