Learn More
ECM Bioscience Myosin IIA Heavy Chain (Ser-1916), phospho-specific Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience MP5191
Encompass_Preferred
This antibody was cross-adsorbed to unphosphorylated Myosin IIA Heavy Chain (Ser-1916) peptide before affinity purification using phospho-Myosin IIA Heavy Chain (Ser-1916) peptide (without carrier). The antibody detects a 200 kDa* protein corresponding to Myosin IIA Heavy Chain on SDS-PAGE immunoblots of human A431 cells treated with Calyculin A. This reactivity is removed after treatment with lambda phosphatase. The immunogen was Phospho-Myosin IIA Heavy Chain (Ser-1916) synthetic peptide (coupled to KLH) corresponding to amino acid residues surrounding Ser-1916 in human myosin IIA heavy chain. This peptide sequence is highly conserved in rat and mouse myosin IIA heavy chain. Accession #P35579. Applications = WB, E; Species Reactivity = Hu, Rt, Ms, Ck; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = NMHC-IIA, MYH9, myosin heavy chain
Catalog No. 50-172-0819