Learn More
AdipoGen anti-Myosin IIA (non-muscle) (heavy chain), mAb (rec.) (SF9) (ATTO 488) (100 µg)
SDP
Supplier:  AdipoGen AG27B0010TDC100
Encompass_Preferred
Recombinant Antibody. Recognizes human, mouse, rat and drosophila myosin IIA (heavy chain). Species cross-reactivity: Drosophila, Human, Mouse, Rat. Clone: SF9. Isotype: Human IgG2lambda. Applications: ICC. Host: Purified from HEK 293 cell culture supernatant. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. anti-Myosin IIA (non-muscle), monoclonal antibody (recombinant) (SF9) is an antibody developed by antibody phage display technology using a human naive antibody gene library.
Catalog No. 50-205-3028