Learn More
CPC Scientific H-Lys-Lys-Arg-Ala-Ala-Arg-Ala-Thr-Ser-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PKCP019A
Encompass
ONE-LETTER SEQUENCE: KKRAARATS-NH2MOLECULAR FORMULA: C40H78N18O11MOLECULAR WEIGHT:987.2STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: R.B. Pearson et al., JBC, 261, 25 (1986)
Catalog No. 50-211-0172