Learn More
Enzo Life Sciences Myosin monoclonal antibody (F109.3E1) (1ml)
SDP
Supplier:  Enzo Life Sciences ALXBC1150SL001
Encompass_Preferred
Clone: F109.3E1. Isotype: IgG1. Immunogen: Both human ventricular myosin light chains. Species reactivity: Human, Rat. Specificity: Recognizes myosin light chain 2 (LC2) (aa 45-59). Applications: IHC (FS), WB. Formulation: Sterile filtered supernatant containing 0.1% sodium azide.
Catalog No. 50-201-0171