Learn More
Medchemexpress LLC Myosmine | 532-12-7 | MFCD00052019 | 146.19 | 250 MG
SDP
Catalog No. 5000450071
Encompass_Preferred
Myosmine is a specific tobacco alkaloid naturally occurring in nuts and nut products. It serves as an analytical standard, primarily intended for research and analytical applications, and is characterized by its low affinity for α4β2 nicotinic acetylcholinergic receptors (nAChR). It has a molecular weight of 146.19 and a purity of greater than 99.7%.
  • A tobacco alkaloid found in natural sources like nuts and nut products
  • Used as an analytical standard for research and analytical applications
  • Exhibits low affinity for α4β2 nicotinic acetylcholinergic receptors
  • Features a molecular weight of 146.19
  • Possesses a purity of greater than 99.7%
Catalog No. 50-004-50071 Supplier Medchemexpress LLC Supplier No. 250MGHYW001909
Myosmine is a specific tobacco alkaloid naturally occurring in nuts and nut products. It serves as an analytical standard, primarily intended for research and analytical applications, and is characterized by its low affinity for α4β2 nicotinic acetylcholinergic receptors (nAChR). It has a molecular weight of 146.19 and a purity of greater than 99.7%.
  • A tobacco alkaloid found in natural sources like nuts and nut products
  • Used as an analytical standard for research and analytical applications
  • Exhibits low affinity for α4β2 nicotinic acetylcholinergic receptors
  • Features a molecular weight of 146.19
  • Possesses a purity of greater than 99.7%