Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cook Myosite MyoTonic Differentiation Medium, 250 mL
SDP

Catalog No. NC2162797
Encompass

The MyoTonic Differentiation Medium is a specialized cell culture medium tailored for the differentiation of myogenic cells, particularly suited for Cook MyoSite's Skeletal Muscle-Derived Cells. This medium supports the critical formation of multi-nucleated myotubes essential for muscle cell research. With decades of experience underpinning its formulation, this medium ensures reliable differentiation and growth of cells.

  • Volume: 250 mL
  • Application: Myogenic cell differentiation, myotube formation
  • Compatibility: Optimized for specific muscle-derived cells, suitable for other myogenic cells
  • Storage temperature: 2-8 C (short-term and long-term)
  • Type: Cell culture medium
  • Usage: Research use only
  • Availability: Non-stock item, manufactured upon request


VENDOR REQUIRES AN INTENDED USE FORM COMPLETED BY END USER

This item is not returnable. View return policy
Catalog No. NC2162797 Supplier Cook Myosite Supplier No. MD55555
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MyoTonic Differentiation Medium is a specialized cell culture medium tailored for the differentiation of myogenic cells, particularly suited for Cook MyoSite's Skeletal Muscle-Derived Cells. This medium supports the critical formation of multi-nucleated myotubes essential for muscle cell research. With decades of experience underpinning its formulation, this medium ensures reliable differentiation and growth of cells.

  • Volume: 250 mL
  • Application: Myogenic cell differentiation, myotube formation
  • Compatibility: Optimized for specific muscle-derived cells, suitable for other myogenic cells
  • Storage temperature: 2-8 C (short-term and long-term)
  • Type: Cell culture medium
  • Usage: Research use only
  • Availability: Non-stock item, manufactured upon request


VENDOR REQUIRES AN INTENDED USE FORM COMPLETED BY END USER

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.