Learn More
Aviva Systems Biology Corp MYOZ1 Antibody - middle region (ARP48398_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48398_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MYOZ1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MYOZ1. Alternate names: CS-2, FATZ, MYOZApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-59813