Learn More
Aviva Systems Biology Corp MYOZ1 Antibody - N-terminal region (ARP48397_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48397_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MYOZ1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human MYOZ1. Alternate names: CS-2, FATZ, MYOZApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-358