Learn More
Alkali Scientific MyPlateFuge™ Microplate Microcentrifuge with Rotor & Plate Carriers, 115v
SDP
Catalog No. 502335850
Encompass_Preferred
The PlateFuge™ centrifuge for microplates is the first 'mini' sized centrifuge with a swing out rotor for microplates. Centrifuging plates before PCR ensures all reactants are in the bottom of the wells for proper concentrations and improved yields.In spite of its remarkably small footprint (9x10in.), the PlateFuge™ includes a uniquely designed, swing out rotor capable of securing 2 microplates. The easily accessible rotor chamber includes two plate carriers that rest at a 75˚ angle. This allows plates to be inserted confidently without sealing tapes or caps. Upon closing the lid of the centrifuge, the rotor accelerates and the centrifugal force "swings' the plates into a horizontal position. Any droplets on the walls of the plate quickly concentrate into the well bottoms.
Catalog No. 50-233-5850 Supplier Alkali Scientific Supplier No. CP2000
Add to Cart
Add to Cart
The PlateFuge™ centrifuge for microplates is the first 'mini' sized centrifuge with a swing out rotor for microplates. Centrifuging plates before PCR ensures all reactants are in the bottom of the wells for proper concentrations and improved yields.In spite of its remarkably small footprint (9x10in.), the PlateFuge™ includes a uniquely designed, swing out rotor capable of securing 2 microplates. The easily accessible rotor chamber includes two plate carriers that rest at a 75˚ angle. This allows plates to be inserted confidently without sealing tapes or caps. Upon closing the lid of the centrifuge, the rotor accelerates and the centrifugal force "swings' the plates into a horizontal position. Any droplets on the walls of the plate quickly concentrate into the well bottoms.