Learn More
Medchemexpress LLC Myr-FEEERA-OH (TFA) | 990.15 | 25 MG
SDP
Catalog No. 5000425842
Encompass_Preferred
mP6 (Myr-FEEERA-OH) is a myristoylated peptide. It inhibits the interaction of Gα13 with integrin β3 without disrupting talin-dependent integrin function, and can block the GTP usage of Rac1, Rap1, and Rab7.
  • Effectively inhibits the infection of CHO-A24 cells
  • Myristoylated peptide
  • Inhibits interaction of Gα13 with integrin β3
  • Blocks GTP usage of Rac1, Rap1, and Rab7
Catalog No. 50-004-25842 Supplier Medchemexpress LLC Supplier No. 25MGHYP10038A
mP6 (Myr-FEEERA-OH) is a myristoylated peptide. It inhibits the interaction of Gα13 with integrin β3 without disrupting talin-dependent integrin function, and can block the GTP usage of Rac1, Rap1, and Rab7.
  • Effectively inhibits the infection of CHO-A24 cells
  • Myristoylated peptide
  • Inhibits interaction of Gα13 with integrin β3
  • Blocks GTP usage of Rac1, Rap1, and Rab7