Learn More
Description
- All Natural
- Food Grade
- FSSC 22000 and ISO 9001 Accreditated Facility
- Additional Food Safety Testing Included (Pesticide, Residual Solvents, Heavy Metals)
Specifications
Specifications
| Chemical Name or Material | Myrcene |
| Packaging | Amber Glass |
| Certificate of Analysis | Yes |
| CAS | 123-35-3 |
| Odor | Balsamic |
| Assay Percent Range | 70% to 100% |
| Physical Form | Liquid |
| Molecular Formula | C10H16 |
| Color | Colorless to Light Yellow |
| Type | Food Additive Standard |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
