Learn More
Enzo Life Sciences Myricetin (10mg). CAS: 529-44-2
SDP
Supplier:  Enzo Life Sciences ALX385012M010
Encompass_Preferred
Antioxidant flavonoid. Has antitumor and chemopreventive properties. Anti-inflammatory. Inhibits NF-κB activation. PIM1 kinase inhibitor IC50=0.78µM. Alternative name: 3,3',4',5,5',7-Hexahydroxyflavone. Purity: ≥90% (HPLC). Appearance: Light yellow-green crystals. Solubility: Soluble in alcohol; slightly soluble in water (warm); insoluble in acetic acid. Long Term Storage: -20°C.
Catalog No. 50-200-9041